D-(+)-Tryptophan

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

7700.1
ROTH7700.1EA 0 EUR
ROTH7700.1
D-(+)-Tryptophan
D-(+)-Tryptophan
Formule: C₁₁H₁₂N₂O₂
Smeltpunt: 280…283 °C
MDL Nummer: MFCD00005647
CAS nummer: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR