Molecular sieve A4 (0.4 nm, 4 Å)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8471.1 8471.2
ROTH8471.1EA 0 EUR
ROTH8471.1 ROTH8471.2
Molecular sieve A4 (0.4 nm, 4 Å)
Molecular sieve A4 (0.4 nm, 4 Å)
Dichtheid: 1,1…2.2 g/cm³ (20 °C)
MDL Nummer: MFCD03457537
CAS nummer: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR