D-(+)-Malic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid

8633.1 8633.2
ROTH8633.1EA 0 EUR
ROTH8633.1 ROTH8633.2
D-(+)-Malic acid
D-(+)-Malic acid
Formule: C₄H₆O₅
MW: 134.09 g/mol
Kookpunt: 322
Smeltpunt: 101…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004245
CAS nummer: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR