Sodium sulphite

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8637.1 8637.2
ROTH8637.1EA 0 EUR
ROTH8637.1 ROTH8637.2
Sodium sulphite
Sodium sulphite
Formule: Na₂SO₃
MW: 126.04 g/mol
Smeltpunt: > 500 °C
Dichtheid: 2.63 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003503
CAS nummer: 7757-83-7
EINECS: 231-821-4
Bereik: 01-2119537420-49
Merck Index: 13,08757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR