L(-)-Malic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

8684.1 8684.2
ROTH8684.1EA 0 EUR
ROTH8684.1 ROTH8684.2
L(-)-Malic acid
L(-)-Malic acid
Formule: C₄H₆O₅
Kookpunt: 322 °C (1013 hPa)
Smeltpunt: 101…104 °C
Dichtheid: 1,595 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064213
CAS nummer: 97-67-6
EINECS: 202-601-5
Merck Index: 13,05730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR