Olive oil

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8873.1 8873.2
ROTH8873.1EA 0 EUR
ROTH8873.1 ROTH8873.2
Olive oil
Olive oil
MDL Nummer: MFCD00131764
CAS nummer: 8001-25-0
EINECS: 232-277-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR