Colchicine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 7α-H-Colchicine, (S)-(-)-Colchicine

8884.1
ROTH8884.1EA 0 EUR
ROTH8884.1
Colchicine
Colchicine
Formule: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Smeltpunt: 154…156 °C
Dichtheid: 1.32 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078484
CAS nummer: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1 I
Merck Index: 12,02536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR