2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9134.1 9134.2 9134.3
ROTH9134.1EA 0 EUR
ROTH9134.1 ROTH9134.2 ROTH9134.3
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formule: C₆H₁₅NO₅S
Kookpunt: 405 °C (1013 hPa)
Smeltpunt: 152…156 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007533
CAS nummer: 10191-18-1
EINECS: 233-465-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR