Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9140.1 9140.2
ROTH9140.1EA 0 EUR
ROTH9140.1 ROTH9140.2
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Formule: C₈H₁₉NO₅
Smeltpunt: 102…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR