Bicine

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9162.1 9162.2
ROTH9162.1EA 0 EUR
ROTH9162.1 ROTH9162.2
Bicine
Bicine
Formule: C₆H₁₃NO₄
Smeltpunt: 185…192 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004295
CAS nummer: 150-25-4
EINECS: 205-755-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR