Stearic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Octadecanoic acid, Octadecylic acid

9459.1 9459.2
ROTH9459.1EA 0 EUR
ROTH9459.1 ROTH9459.2
Stearic acid
Stearic acid
Formule: C₁₈H₃₆O₂
MW: 284.48 g/mol
Kookpunt: 361
Smeltpunt: 67
Dichtheid: 0.94 g/cm³ (20 °C)
Vlampunt: 196 °C (gesloten beker)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002752
CAS nummer: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR