Zetmeel, hydrolysed acc. Smithies

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9480.1 9480.2 9480.4
ROTH9480.1EA 0 EUR
ROTH9480.1 ROTH9480.2 ROTH9480.4
Zetmeel, hydrolysed acc. Smithies
Zetmeel
Formule: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082026
CAS nummer: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR