N,N-Dimethylformamide

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A529.1 A529.2
ROTHA529.1EA 151.5 EUR
ROTHA529.1 ROTHA529.2
N,N-Dimethylformamide
N,N-Dimethylformamide
Formule: HCON(CH₃)₂
Kookpunt: 153 °C (1013 hPa)
Smeltpunt: –61 °C
Dichtheid: 0,949 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
Bereik: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR