Dichloromethane-[D₂]

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

AE55.1 AE55.2
ROTHAE55.1EA 142.5 EUR
ROTHAE55.1 ROTHAE55.2
Dichloromethane-[D₂]
Dichloromethane-[D₂]
Formule: CD₂Cl₂
Kookpunt: 40 °C (1013 hPa)
Smeltpunt: –97 °C
Dichtheid: 1,36 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000882
CAS nummer: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR