Ampicillin sodium salt

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

K029.1 K029.2
ROTHK029.1EA 28.5 EUR
ROTHK029.1 ROTHK029.2
Ampicillin sodium salt
Ampicillin sodium salt
Formule: C₁₆H₁₈N₃NaO₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00064313
CAS nummer: 69-52-3
EINECS: 200-708-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR