Calcium carbonate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Marble

P013.1 P013.2
ROTHP013.1EA 19.9 EUR
ROTHP013.1 ROTHP013.2
Calcium carbonate
Calcium carbonate
Formule: CaCO₃
MW: 100.09 g/mol
Smeltpunt: 825
Dichtheid: 2.71 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010906
CAS nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR