Ammonium iron(III) sulphate dodecahydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate

P734.1 P734.2
ROTHP734.1EA 85.9 EUR
ROTHP734.1 ROTHP734.2
Ammonium iron(III) sulphate dodecahydrate
Ammonium iron(III) sulphate dodecahydrate
Formule: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Kookpunt: 230
Smeltpunt: 39…41 °C
Dichtheid: 1.71 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150004
CAS nummer: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR