Fuchsin (acid)

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: CI 42685

T128.1
ROTHT128.1EA 49.5 EUR
ROTHT128.1
Fuchsin (acid)
Fuchsin (acid)
Formule: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Smeltpunt: > 130 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013286
CAS nummer: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR