Water

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T143.1 T143.2 T143.3 T143.4 T143.5
ROTHT143.1EA 30.5 EUR
ROTHT143.1 ROTHT143.2 ROTHT143.3 ROTHT143.4 ROTHT143.5
Water
Water
Formule: H₂O
Kookpunt: 100 °C (1013 hPa)
Smeltpunt: 0 °C
Dichtheid: 1 g/cm³ (4 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR