Calcium chloride dihydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Calcium Chloride Dihydrate, Calcium dichloride

T885.1 T885.2 T885.3
ROTHT885.1EA 37.9 EUR
ROTHT885.1 ROTHT885.2 ROTHT885.3
Calcium chloride dihydrate
Calcium chloride dihydrate
Formule: CaCl₂·2H₂O
MW: 147.01 g/mol
Kookpunt: 1600
Smeltpunt: 176
Dichtheid: 1.85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149613
CAS nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR