Cobalt(II) chloride hexahydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

T889.1
ROTHT889.1EA 219 EUR
ROTHT889.1
Cobalt(II) chloride hexahydrate
Cobalt(II) chloride hexahydrate
Formule: CoCl₂·6H₂O
MW: 237.93 g/mol
Kookpunt: 110
Smeltpunt: 56
Dichtheid: 1.92 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149652
CAS nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 13,02462

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR