2-Propanol

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

T910.1
ROTHT910.1EA 93.9 EUR
ROTHT910.1
2-Propanol
2-Propanol
Formule: (CH₃)₂CHOH
Kookpunt: 82 °C (1013 hPa)
Smeltpunt: –89 °C
Dichtheid: 0,786 g/cm³ (20 °C)
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011674
CAS nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
Bereik: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR