Zinc acetate dihydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

X893.1 X893.2
ROTHX893.1EA 30.5 EUR
ROTHX893.1 ROTHX893.2
Zinc acetate dihydrate
Zinc acetate dihydrate
Formule: (H₃CCOO)₂Zn·2H₂O
Smeltpunt: 237 °C
Dichtheid: 1,718 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00066961
CAS nummer: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR