Ureum

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

X999.1 X999.2 X999.3
ROTHX999.1EA 18.9 EUR
ROTHX999.1 ROTHX999.2 ROTHX999.3
Ureum
Ureum
Formule: NH₂CONH₂
Kookpunt: 197 °C (1013 hPa)
Dichtheid: 1,34 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
Bereik: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR