Respirators, T200 Series
Leverancier: RPB SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17-200-22-CE 17-200-32-CE 04-852 17-722 17-732 17-712 17-811 17-815 17-817 15-860 17-513 04-831 04-841 16-526
RPBS17-200-22-CEEA 0 EUR
RPBS17-200-22-CE RPBS17-200-32-CE RPBS04-852 RPBS17-722 RPBS17-732 RPBS17-712 RPBS17-811 RPBS17-815 RPBS17-817 RPBS15-860 RPBS17-513 RPBS04-831 RPBS04-841 RPBS16-526
Respirators, T200 Series
Ademhalingsmaskers Ademhalingssystemen
The RPB T200 is a lightweight multi-purpose respirator designed specifically for the healthcare and life science industries, relieving workers of breathing difficulties and fatigue experienced with N95 use. Available with either a head harness or bump cap for head protection and multiple hood options, the T200 has the ability to adapt to different environments that need face protection and respiratory protection.
  • Customizable internal layout with four adjustable head harness points and moveable lens and air duct system
  • Deconable head suspension system and
More Product Information
Order Now
Specificaties
Environmentally Preferable