Bekertangen
Leverancier: RSG
Ratings:
Avg. Ratings: 4.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
043.250 043.220 043.200 043.300
RSGA043.250EA 30.3 EUR
RSGA043.250 RSGA043.220 RSGA043.200 RSGA043.300
Bekertangen
Tangen
These beaker/cup tongs are made from electrolytically polished stainless steel.
  • Jaws with cork lining