Special tweezers, coated
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
416.110 416.130
RSGA416.110EA 129 EUR
RSGA416.110 RSGA416.130
Special tweezers, coated
Pincetten
These special tweezers made of stainless steel feature a dark PTFE coating.
  • Extraordinarily fine
  • Precision cut tips