Foetal bovine serum (FBS), USDA approved
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S1700-100 S1700-500 S1650-100 S1650-500 S1600-500 S1560-100 S1560-500 S156H-100 S156H-500 S165H-500 S1600-100 S165H-100 S160H-100 S160H-500 S170G-100 S170G-500 S170H-100 S170H-500 S165G-500 S160B-100 S160B-500
S1700-100EA 0 EUR
S1700-100 S1700-500 S1650-100 S1650-500 S1600-500 S1560-100 S1560-500 S156H-100 S156H-500 S165H-500 S1600-100 S165H-100 S160H-100 S160H-500 S170G-100 S170G-500 S170H-100 S170H-500 S165G-500 S160B-100 S160B-500
Foetal bovine serum (FBS), USDA approved
Serum
Sterile filtered foetal bovine serum (FBS) derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • High quality serum, rigorously tested
  • Wide range of sources available
  • Full traceability and security
USDA approved sources from countries where excellent veterinary conditions are available. EDQM certified FBS guarantees that the origin and the manufacturing process is certified by the European Directorate for theMore Product Information