Mouse serum
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S2160-020 S2160-050 S2160-100 S2160-500 S2160-010
S2160-020EA 101 EUR
S2160-020 S2160-050 S2160-100 S2160-500 S2160-010
Mouse serum
Serum
Mouse serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Mouse serum is used as a blocking agent in immunoassays and controls.