Samplers for cohesive powders
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2015A-05 2015A-25 2015A-10
SAMP2015A-05EA 1450 EUR
SAMP2015A-05 SAMP2015A-25 SAMP2015A-10
Samplers for cohesive powders
Monsternemers Monsternemers voor vaste stoffen
These samplers are meant to work from a known point within the product.
Suitable for sticky cohesive powders.
Leveringsinformatie: Supplied as standard with pointed tip.