Pump sampler sets
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5020S-02
SAMP5020S-02EA 1780 EUR
SAMP5020S-02
Pump sampler sets
Monsternemers Monsternemers voor vloeistoffen
These pump samplers allow for the quick sampling of liquids.
  • Reduced contamination by using new tube and bottles for each sample
  • Only the tube and collecting bottle come into contact with the products
  • High efficiency and ability to pump water to a height of about four meters
Suitable for low to medium viscosity liquids.