Powder funnels, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8111H-200 8111H-200S 8111P-350S 8111P-350 8111H-100 8111H-100S 8111H-50 8111H-50S
SAMP8111H-200EA 102 EUR
SAMP8111H-200 SAMP8111H-200S 300-0533 300-0532 SAMP8111H-100 SAMP8111H-100S SAMP8111H-50 SAMP8111H-50S
Powder funnels, SteriWare®
Trechters Poedertrechters
These single use powder funnels made from chemically resistant, food grade HDPE are ideal for use with powders, creams and liquids.
  • Available as sterile and non sterile versions
  • Manufactured under cleanroom conditions: Can be used straight from the bag
  • Cross-contamination risk is eliminated
  • No cleaning is required before or after sampling
The wide opening at the base of the funnel will help prevent powder blockages.
Certificaten: Material conforms to FDC and EC standards. Full batch traceability.
Verpakking: Individually bagged.