SteriWare Multi Grip Tweezers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8270A-200 8270A-200S 8270A-300 8270A-300S
SAMP8270A-200EA 160 EUR
SAMP8270A-200 SAMP8270A-200S SAMP8270A-300 SAMP8270A-300S
SteriWare Multi Grip Tweezers
Pincetten
These SteriWare multi grip tweezers operate like conventional tweezers, they have an additional grip feature enabling the easy and secure handling of small vials and containers.
  • Made from food grade PS
  • Individually bagged to prevent cross contamination
  • 100% manufactured and packed in a cleanroom
  • Choice of sterile and non sterile