X-ray and metal detectable pallet knife and stirrer
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M1016B-200
SAMPM1016B-200EA 31.5 EUR
SAMPM1016B-200
X-ray and metal detectable pallet knife and stirrer
Spatels
The X-ray and metal detectable pallet knife is ideal for scraping and mixing material. The pallet knives are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Long blade, can be used to stir and mix containers
  • Full batch traceability
  • The 200 mm blade can be used to stir and mix containers
To ensure the cleanliness of the pallet knives are manufacturedMore Product Information