Wire weight, OIML F1
Leverancier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
YCW1131-00
SARBYCW1131-00EA 43.5 EUR
SARBYCW1131-00
Wire weight, OIML F1
Gewichten Kalibratiegewichten
Accurate metrological weight with nominal mass value of 10 mg.
  • OIML class F1
  • Hardness: ca. 110 HB
  • Density of 7,950 kg/m³
  • High gloss polished surface
  • No laser markings
  • Without certificate
Certificaten: Weights comply with the International Recommendation OIML-R111.