Calibration Weights, OIML, M1
Leverancier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
YCW355-AC-02 YCW325-AC-02 YCW315-AC-02 YCW315-AC-00 YCW325-AC-00 YCW625-AC-02 YCW625-AC-00 YCW355-AC-00 YCW615-AC-00 YCW555-AC-02 YCW555-AC-00 YCW615-AC-02 YCW715-AC-02 YCW715-AC-00 YCW655-AC-02 YCW655-AC-00 YCW425-AC-02 YCW425-AC-00 YCW455-AC-00 YCW415-AC-02 YCW415-AC-00 YCW525-AC-02 YCW525-AC-00 YCW515-AC-02 YCW515-AC-00 YCW455-AC-02
SARBYCW355-AC-02EA 79.6 EUR
SARBYCW355-AC-02 SARBYCW325-AC-02 SARBYCW315-AC-02 SARBYCW315-AC-00 SARBYCW325-AC-00 SARBYCW625-AC-02 SARBYCW625-AC-00 SARBYCW355-AC-00 SARBYCW615-AC-00 SARBYCW555-AC-02 SARBYCW555-AC-00 SARBYCW615-AC-02 SARBYCW715-AC-02 SARBYCW715-AC-00 SARBYCW655-AC-02 SARBYCW655-AC-00 SARBYCW425-AC-02 SARBYCW425-AC-00 SARBYCW455-AC-00 SARBYCW415-AC-02 SARBYCW415-AC-00 SARBYCW525-AC-02 SARBYCW525-AC-00 SARBYCW515-AC-02 SARBYCW515-AC-00 SARBYCW455-AC-02
Calibration Weights, OIML, M1
Gewichten Kalibratiegewichten
Weights are made from polished brass.
Leveringsinformatie: Supplied in a plastic screw-top box.