Filtration capsules, Sartopore® 2
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5445307H7-SS-A 5445306G7-OO-A 5445307H7-SO-A 5445307H9-SO-A 5445307H8-SO-A 5445358K7-SO-A 5445307HS-HH-M 5441307H4G-OOB 5445307H8G-OOA 5445307H9-FF-A 5445307H7-FF-A 5445307H0-FF-V 5445307G7-FF-A
SARP5445307H7-SS-AEA 544 EUR
SARP5445307H7-SS-A SARP5445306G7-OO-A SARP5445307H7-SO-A SARP5445307H9-SO-A SARP5445307H8-SO-A SARP5445358K7-SO-A SARP5445307HS-HH-M SARP5441307H4G-OOB SARP5445307H8G-OOA SARP5445307H9-FF-A SARP5445307H7-FF-A SARP5445307H0-FF-V SARP5445307G7-FF-A
Filtration capsules, Sartopore® 2
Filters Membraanfiltercapsules
Ready to use disposable capsules for sterile filtration in laboratory scale or for small scale filterability trials.
  • Ideal for scale-up and scale-down
  • Compatible with a pH range from pH one to 14
  • Typical applications include sterilising grade filtration of therapeutics, biological fluids, injectables, purified water, media, buffers
Hydrophilic PES membrane, PP housing.
Each individual element is tested for integrity by bubble point and diffusion test prior to be released assuring absolute reliability.