Blood collection set, Minivette®
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17.2112.050
SARS17.2112.050EA 0 EUR
SARS17.2112.050
Blood collection set, Minivette®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection set for the precise dispensing of small volumes.
  • Easy sample recovery
  • Hygienic and secure sample transfer between patient and test equipment
Used in highly infectious nursing sectors.
Verpakking: Individually wrapped.
Order Now
Specificaties
Environmentally Preferable