Reagent/centrifuge tubes, skirted base, with screw cap
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60.732.001 60.558.001 60.541.685 62.559.001 60.9922.243 60.549.001 60.732 62.543 60.540.686 60.549 60.551 60.551.001 60.558 60.9921.821 60.9921.829 60.9921.830 60.595.241 62.551.201 62.559
SARS60.732.001EA 102 EUR
SARS60.732.001 SARS60.558.001 SARS60.541.685 SARS62.559.001 SARS60.9922.243 SARS60.549.001 SARS60.732 SARS62.543 SARS60.540.686 SARS60.549 SARS60.551 SARS60.551.001 SARS60.558 SARS60.9921.821 SARS60.9921.829 SARS60.9921.830 SARS60.595.241 SARS62.551.201 SARS62.559
Reagent/centrifuge tubes, skirted base, with screw cap
Buisjes Centrifugebuisjes
PP, translucent oder PS, transparent with HDPE screw caps
  • Non sterile or sterile
  • Caps enclosed or assembled available
  • With or without graduation and writing space
Universal tubes for a wide range of applications.