Micro tubes with thread
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
72.693.465 72.730.217
SARS72.693.465EA 174.48 EUR
SARS72.693.465 SARS72.730.217
Micro tubes with thread
Buisjes Microbuisjes
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.