Microcentrifuge tubes, SafeSeal
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
72.706 72.706.200 72.695.001 72.695.002 72.695.004 72.695.005 72.695.006 72.695.007 72.695.200 72.704 72.704.002 72.706.002 72.704.004 72.704.005 72.704.006 72.695.500
SARS72.706EA 63.18 EUR
SARS72.706 SARS72.706.200 SARS72.695.001 SARS72.695.002 SARS72.695.004 SARS72.695.005 SARS72.695.006 SARS72.695.007 SARS72.695.200 SARS72.704 SARS72.704.002 SARS72.706.002 SARS72.704.004 SARS72.704.005 SARS72.704.006 SARS72.695.500
Microcentrifuge tubes, SafeSeal
Buisjes Microbuisjes
PP, translucent.
  • Wide hinge for easy and precise closure
  • Large writing space on lid, graduated
  • Centrifugation to 30000×g (2,0 ml tubes: to 25000×g)
For transport, storage and sample processing in the laboratory. DNA-/RNase-free tubes and screw caps for the storage of high-grade synthesised oligonucleotides, enzymes, buffers etc. Retaining cams ensure a safe fit also under thermal strain.