Faeces containers
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
80.734.301 80.620 80.621 80.622 80.622.111 80.623 80.623.111 80.734 80.734.001 80.734.311 80.734.401
SARS80.734.301EA 102.75 EUR
SARS80.734.301 SARS80.620 SARS80.621 SARS80.622 SARS80.622.111 SARS80.623 SARS80.623.111 SARS80.734 SARS80.734.001 SARS80.734.311 SARS80.734.401
Faeces containers
Diagnostische systemen Clinical Sample Collection/Preparation Faeces Sample Collection
Containers with sampling scoops enables clean and easy collection of faeces samples.
  • To collect defined faeces volume of 1 ml, approximately 1 g
  • Sterile or non sterile
  • Labelled or opaque options available