Deep well plates, 96-well, PP, MegaBlock®
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
82.1971.002 82.1972.002
SARS82.1971.002EA 134.93 EUR
SARS82.1971.002 SARS82.1972.002
Deep well plates, 96-well, PP, MegaBlock®
Microplaten Deepwell-microplaten
PP. The 96-well, 1,2/2,2 ml MegaBlock® meets all important requirements for automated sample processing or storing large quantities of backup samples.
  • Each well 100% leakproof tested
  • Suitable for DNA isolation, enzyme assays and tissue culture applications
  • Ideal for longterm storage of pharmaceutical samples
  • Solvent resistant to DMSO
  • Round bottom wells
  • Alphanumeric coding for easy identification