Multi-tube racks, S-Monovette®
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
93.837 93.826 93.850 93.841.100 93.1431.001 93.838 93.848.100 93.894 93.1097.100 93.1431 93.48.011 93.827 93.827.013 93.838.013 93.844.100 93.846.100 93.849.100 93.850.001 93.868 93.870 93.893.100 93.1428
SARS93.837EA 37.79 EUR
SARS93.837 SARS93.826 SARS93.850 SARS93.841.100 SARS93.1431.001 SARS93.838 SARS93.848.100 SARS93.894 SARS93.1097.100 SARS93.1431 SARS93.48.011 SARS93.827 SARS93.827.013 SARS93.838.013 SARS93.844.100 SARS93.846.100 SARS93.849.100 SARS93.850.001 SARS93.868 SARS93.870 SARS93.893.100 SARS93.1428
Multi-tube racks, S-Monovette®
Rekken Tube Racks
Made from unbreakable, transparent plastic and can be easily disassembled for cleaning.
  • A wide range with a variety of diameters
  • High-grade and resistant material that is durable
  • Available in different colours
These universal compact block rack are ideal for space-saving and systematic sample storage.