Transport Boxes
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95.1715 95.1717 95.1717.003 95.1717.011
SARS95.1715EA 316.07 EUR
SARS95.1715 SARS95.1717 SARS95.1717.003 SARS95.1717.011
Transport Boxes
Dozen Transportdozen
Transport case housing is manufactured from durable polyethylene and offers optimum protection for primary receptacles during transport.
  • Economical, flexible
  • Central one-hand closure
  • Lateral tabs for easy attachment of straps or restraints
  • Transparent window label for easy identification
Transport cases are suitable for transportation of diagnostic samples and blood products.
Certificaten: Complies with the requirement of P650 packaging instructions for class UN3373 of the ADR and RID.