Syringe filter holder, stainless steel
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16214
SART16214EA 267 EUR
SART16214
Syringe filter holder, stainless steel
Filterhouders
This syringe filter holder is made from stainless steel. PTFE coated surface on the top provides leak proof sealing, and the chemical compatibility depends upon the filter inserted.
  • Re-usable
  • Resistant to heat