Vacuum hand pump
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16673
SART16673EA 418 EUR
SART16673
Vacuum hand pump
Pompen Vacuümhandpompen
The hand operated vacuum pump made of PVC is used as practical vacuum source for outside of a laboratory.
Leveringsinformatie: Supplied completely with gauge, vacuum release lever and a clear plastic tubing (600 mm).