Test tubes without rim, soda lime glass
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A402507000711 A415016000711 A405511000811 A405006500711 A404010500911 A407512000611 A407510000611 A406012000611 A406512000611 A408512000711 A412012000711 A416018001011 A412516000711 A405009500811 A403506500711 A404009500711 A406514500811 A410018001011 A416017001011 A415015750911 A401705000711
SCERA402507000711EA 347 EUR
SCERA402507000711 SCERA415016000711 SCERA405511000811 SCERA405006500711 SCERA404010500911 SCERA407512000611 SCERA407510000611 SCERA406012000611 SCERA406512000611 SCERA408512000711 SCERA412012000711 SCERA416018001011 SCERA412516000711 SCERA405009500811 SCERA403506500711 SCERA404009500711 SCERA406514500811 SCERA410018001011 SCERA416017001011 SCERA415015750911 SCERA401705000711
Test tubes without rim, soda lime glass
Buisjes Reageerbuisjes
Use the soda lime glass culture tube to view cell cultures in the laboratory.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable