Tissue grinders, CS1, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP749600-0015 SP749600-0050
SCERSP749600-0015EA 267 EUR
SCERSP749600-0015 SCERSP749600-0050
Tissue grinders, CS1, KIMBLE®
Homogenisatoren Potterbuizen
CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
  • Narrow pestles eliminate the potential of sample overflow
  • Convenient and easy-to-use
  • Gamma sterilised in an easy-open Tyvek® pack
The molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination. Tubes are made of PP, pestles are 30% glass-filled PP and caps are HDPE.
Leveringsinformatie: Each tissue grinder is supplied with a fully assembled pestle, a conical sample tubeMore Product Information