Tissue grinders, DUALL®, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP885480-0020 SP885480-0022 SP885480-0023
SCERSP885480-0020EA 228 EUR
SCERSP885480-0020 SCERSP885480-0022 SCERSP885480-0023
Tissue grinders, DUALL®, KIMBLE®
Homogenisatoren Potterbuizen
These tissue grinders (borosilicate glass, PTFE pestle with glass tube) combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Construction is strong enough to allow the pestle to be motor driven and also the grinding efficiency is greatly improved while using this tube.
  • Replacement components are available and completely interchangeable
  • Used for soft tissue such as brain and liver
Clearance between pestles and tubes is 0,004 to 0,006"